Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Brpf3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081315.1 | NP_001074784.1 | 10 - 11 | 3438 | 70 | |
GenBank mRNA | AK129322.1 | - | 8 - 9 | 1693 | 70 | |
AK147555.1 | - | 10 - 11 | 3437 | 70 | ||
AK162638.1 | - | 3 - 4 | 286 | 70 | ||
BC157915.1 | - | 9 - 10 | 3203 | 9088158 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |