Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Jmjd8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_028101.4 | NP_082377.2 | 3 - 4 | 271 | 66 | |
GenBank mRNA | AK011308.1 | - | 3 - 4 | 268 | 66 | |
AK148079.1 | - | 3 - 4 | 249 | 66 | ||
AK155810.1 | - | 3 - 4 | 273 | 66 | ||
AK172073.1 | - | 3 - 4 | 268 | 66 | ||
BC010800.1 | - | 3 - 4 | 233 | 4235072 | 66 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |