Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Chtf18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_145409.2 | NP_663384.2 | 11 - 12 | 1539 | 62 | |
GenBank mRNA | AK052673.1 | - | 11 - 12 | 1473 | 62 | |
AK077947.1 | - | 11 - 12 | 1506 | 62 | ||
BC024142.1 | - | 11 - 12 | 1526 | 5149957 | 62 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |