Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fbxl16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001164225.1 | NP_001157697.1 | 4 - 5 | 1543 | 71 | |
GenBank mRNA | BC042620.1 | - | 3 - 4 | 1252 | 71 | |
BC132383.1 | - | 3 - 4 | 1339 | 40130660 | 71 | |
BC137657.1 | - | 3 - 4 | 1339 | 8860676 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |