Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tex13a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026469.2 | NP_080745.2 | 1 - 2 | 612 | 77 | |
GenBank mRNA | AK006452.1 | - | 1 - 2 | 611 | 77 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Detect Genomic DNA Mus musculus 1700 Probe spans exons |