Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mslnl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_177822.3 | NP_808490.1 | 11 - 12 | 1586 | 57 | |
GenBank mRNA | AK028891.1 | - | 11 - 12 | 1586 | 57 | |
BC052484.1 | - | 10 - 11 | 1322 | 6312790 | 57 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |