Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wnt7b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001163633.1 | NP_001157105.1 | 3 - 4 | 980 | 77 | |
NM_001163634.1 | NP_001157106.1 | 3 - 4 | 1064 | 77 | ||
NM_009528.3 | NP_033554.3 | 3 - 4 | 943 | 77 | ||
GenBank mRNA | AK087499.1 | - | 3 - 4 | 981 | 77 | |
BC052018.1 | - | 3 - 4 | 704 | 6401357 | 77 | |
BC058398.1 | - | 3 - 4 | 939 | 77 | ||
BC066003.1 | - | 3 - 4 | 1064 | 5701652 | 77 | |
M89802.1 | - | 3 - 4 | 572 | 77 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Mus musculus Inventoried Probe spans exons |