Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Upk3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_023478.2 | NP_075967.2 | 3 - 4 | 565 | 71 | |
GenBank mRNA | AK003737.1 | - | 1 - 2 | 76 | 71 | |
BC125336.1 | - | 3 - 4 | 565 | 40129851 | 71 | |
BC125338.1 | - | 3 - 4 | 565 | 40129853 | 71 | |
BX512009.1 | - | 2 - 3 | 372 | 71 | ||
BY703997.1 | - | 1 - 2 | 76 | 71 | ||
DQ387454.1 | - | 3 - 4 | 515 | 71 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |