Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Erbb2ip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001005868.2 | NP_001005868.1 | 11 - 12 | 1289 | 153 | |
NM_001289473.1 | NP_001276402.1 | 11 - 12 | 1289 | 153 | ||
NM_001289474.1 | NP_001276403.1 | 11 - 12 | 1289 | 153 | ||
NM_001289475.1 | NP_001276404.1 | 11 - 12 | 1289 | 153 | ||
GenBank mRNA | AK122473.1 | - | 11 - 12 | 968 | 153 | |
AK169529.1 | - | 11 - 12 | 1218 | 153 | ||
BB376299.2 | - | 5 - 6 | 483 | 153 | ||
BC141178.1 | - | 11 - 12 | 998 | 9055790 | 153 | |
BC141188.1 | - | 11 - 12 | 998 | 9055800 | 153 | |
BC145498.1 | - | 11 - 12 | 998 | 9054061 | 153 | |
BC145509.1 | - | 11 - 12 | 998 | 9054072 | 153 | |
EU233994.1 | - | 13 - 14 | 2046 | 153 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |