Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ecel1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001277925.1 | NP_001264854.1 | 8 - 9 | 1720 | 72 | |
NM_021306.3 | NP_067281.2 | 8 - 9 | 1712 | 72 | ||
GenBank mRNA | AB026294.1 | - | 7 - 8 | 1540 | 72 | |
AK039467.1 | - | 8 - 9 | 1713 | 72 | ||
AK159365.1 | - | 8 - 9 | 1722 | 72 | ||
BC057569.1 | - | 7 - 8 | 1542 | 6413731 | 72 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Protease Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |