Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Col1a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_007742.3 | NP_031768.2 | 4 - 5 | 437 | 123 | |
GenBank mRNA | AK132180.1 | - | 4 - 5 | 453 | 123 | |
AK158011.1 | - | 4 - 5 | 453 | 123 | ||
AK159283.1 | - | 4 - 5 | 455 | 123 | ||
AK159285.1 | - | 4 - 5 | 457 | 123 | ||
BC050014.1 | - | 4 - 5 | 437 | 4225038 | 123 | |
BC059281.1 | - | 4 - 5 | 447 | 3992363 | 123 | |
U08020.1 | - | 4 - 5 | 338 | 123 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |