Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dennd2c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_177857.1 | NP_808525.2 | 2 - 3 | 1067 | 80 | |
GenBank mRNA | AK044389.1 | - | 2 - 3 | 1050 | 80 | |
BC060666.1 | - | 2 - 3 | 1067 | 6841982 | 80 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |