Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ddx52 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_030096.2 | NP_084372.2 | 1 - 2 | 170 | 76 | |
GenBank mRNA | AK019204.1 | - | 1 - 2 | 93 | 76 | |
AK080767.1 | - | 1 - 2 | 108 | 76 | ||
AK133545.1 | - | 1 - 2 | 141 | 76 | ||
AK141200.1 | - | 1 - 2 | 144 | 76 | ||
AK166789.1 | - | 1 - 2 | 96 | 76 | ||
BC029094.1 | - | 1 - 2 | 82 | 3662249 | 76 | |
CJ142321.1 | - | 1 - 2 | 171 | 76 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus 1700 Probe spans exons |