Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Amdhd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172935.4 | NP_766523.2 | 1 - 2 | 157 | 86 | |
GenBank mRNA | AK077584.1 | - | 1 - 2 | 140 | 86 | |
AK090009.1 | - | 1 - 2 | 159 | 86 | ||
AK154209.1 | - | 1 - 2 | 157 | 86 | ||
AK159860.1 | - | 1 - 2 | 154 | 86 | ||
AK168432.1 | - | 1 - 2 | 115 | 86 | ||
BC037005.1 | - | 1 - 2 | 193 | 5101453 | 86 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Probe spans exons |