Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tgfb3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009368.3 | NP_033394.2 | 1 - 2 | 1462 | 130 | |
GenBank mRNA | AK132519.1 | - | 1 - 2 | 1452 | 130 | |
AK145304.1 | - | 1 - 2 | 1447 | 130 | ||
AK148385.1 | - | 1 - 2 | 1326 | 130 | ||
AK162560.1 | - | 1 - 2 | 226 | 130 | ||
BC005513.1 | - | 1 - 2 | 204 | 130 | ||
BC014690.1 | - | 1 - 2 | 741 | 3663150 | 130 | |
BC089347.1 | - | 1 - 2 | 1419 | 130 | ||
BC094591.1 | - | 2 - 3 | 887 | 130 | ||
BC108426.1 | - | 1 - 2 | 1462 | 6757707 | 130 | |
M32745.1 | - | 1 - 2 | 957 | 130 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus 1700 Probe spans exons |