Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lgals9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001159301.1 | NP_001152773.1 | 8 - 9 | 773 | 100 | |
NM_010708.2 | NP_034838.2 | 9 - 10 | 866 | 100 | ||
GenBank mRNA | AK146044.1 | - | 8 - 9 | 729 | 100 | |
AK214388.1 | - | 3 - 4 | 164 | 100 | ||
BC003754.1 | - | 8 - 9 | 700 | 3601419 | 100 | |
U55060.1 | - | 8 - 9 | 699 | 100 | ||
U55061.1 | - | 9 - 10 | 773 | 100 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Apoptosis Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |