Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ift20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_018854.4 | NP_061342.1 | 2 - 3 | 368 | 58 | |
GenBank mRNA | AK002397.1 | - | 2 - 3 | 369 | 58 | |
AK139942.1 | - | 2 - 3 | 405 | 58 | ||
AK213963.1 | - | 1 - 2 | 238 | 58 | ||
BC002040.1 | - | 2 - 3 | 359 | 3590790 | 58 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |