Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lig3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001291245.1 | NP_001278174.1 | 4 - 5 | 1066 | 78 | |
NM_001291246.1 | NP_001278175.1 | 4 - 5 | 1063 | 78 | ||
NM_001291247.1 | NP_001278176.1 | 4 - 5 | 1067 | 78 | ||
NM_010716.3 | NP_034846.2 | 4 - 5 | 1070 | 78 | ||
GenBank mRNA | AK031540.1 | - | 4 - 5 | 1008 | 78 | |
AK143531.1 | - | 4 - 5 | 936 | 78 | ||
AK150644.1 | - | 4 - 5 | 975 | 78 | ||
AK156018.1 | - | 4 - 5 | 985 | 78 | ||
AK167363.1 | - | 4 - 5 | 978 | 78 | ||
BC026143.1 | - | 4 - 5 | 1038 | 78 | ||
BC049240.1 | - | 4 - 5 | 973 | 4504384 | 78 | |
BC083500.1 | - | 4 - 5 | 969 | 3673103 | 78 | |
U66057.1 | - | 4 - 5 | 1004 | 87 | ||
U66058.1 | - | 4 - 5 | 1004 | 87 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |