Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mdga1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081160.1 | NP_001074629.1 | 14 - 15 | 3013 | 71 | |
GenBank mRNA | AK004706.1 | - | 7 - 8 | 1023 | 71 | |
DQ788983.1 | - | 3442 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Mus musculus Probe spans exons |