Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1100011711.mm.1 |
NM_001035854.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100011711.mm.1 |
NM_001035854.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100011711.mm.1 |
NM_027915.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100011711.mm.1 |
NM_027915.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100011711.mm.1 |
TC1100001247.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100011715.mm.1 |
NM_001035854.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100011715.mm.1 |
NM_001035854.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100011715.mm.1 |
NM_027915.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100011715.mm.1 |
NM_027915.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100011715.mm.1 |
TC1100001247.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100011707.mm.1 |
NM_001035854.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100011707.mm.1 |
NM_001035854.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100011707.mm.1 |
NM_027915.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100011707.mm.1 |
NM_027915.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100011707.mm.1 |
TC1100001247.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100011712.mm.1 |
NM_001035854.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100011712.mm.1 |
NM_001035854.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100011712.mm.1 |
NM_027915.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100011712.mm.1 |
NM_027915.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100011712.mm.1 |
TC1100001247.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100011714.mm.1 |
NM_001035854.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100011714.mm.1 |
NM_001035854.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100011714.mm.1 |
NM_027915.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100011714.mm.1 |
NM_027915.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100011714.mm.1 |
TC1100001247.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100011706.mm.1 |
NM_001035854.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100011706.mm.1 |
NM_001035854.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100011706.mm.1 |
NM_027915.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100011706.mm.1 |
NM_027915.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100011706.mm.1 |
TC1100001247.mm.1 |
MTA TC |