Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zcrb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK012589.1 | - | 3 - 4 | 262 | 128 |
Set Membership: |
Amplicon length greater than or equal to 101 Mus musculus Probe spans exons |