Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cpne8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_025815.2 | NP_080091.1 | 3 - 4 | 306 | 180 | |
GenBank mRNA | AK004559.1 | - | 3 - 4 | 294 | 180 | |
AK010227.1 | - | 3 - 4 | 306 | 180 | ||
AK013407.1 | - | 3 - 4 | 291 | 180 | ||
BC017626.1 | - | 3 - 4 | 277 | 4502073 | 180 | |
BC138745.1 | - | 3 - 4 | 305 | 8861767 | 180 | |
BC138746.1 | - | 3 - 4 | 305 | 8861768 | 180 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |