Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Smgc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_198927.3 | NP_945121.1 | 2 - 3 | 228 | 79 | |
GenBank mRNA | AK017450.1 | - | 2 - 3 | 229 | 79 | |
AK132579.1 | - | 2 - 3 | 230 | 79 | ||
AY459348.1 | - | 2 - 3 | 229 | 79 | ||
BC055490.1 | - | 2 - 3 | 79 | |||
BC092073.1 | - | 1 - 2 | 79 | |||
BC138091.1 | - | 2 - 3 | 8861110 | 79 | ||
BC145136.1 | - | 2 - 3 | 229 | 9053684 | 79 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |