Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Agxt2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK149538.1 | - | 7 - 8 | 792 | 106 | |
BC141248.1 | - | 7 - 8 | 805 | 9055860 | 106 | |
BC145576.1 | - | 7 - 8 | 802 | 9054142 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |