Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Suz12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001163018.1 | NP_001156490.1 | 13 - 14 | 1937 | 96 | |
NM_199196.2 | NP_954666.1 | 14 - 15 | 2006 | 96 | ||
GenBank mRNA | AK141264.1 | - | 5 - 6 | 610 | 96 | |
BC039915.1 | - | 8 - 9 | 1082 | 96 | ||
BC051099.1 | - | 5 - 6 | 700 | 96 | ||
BC064461.1 | - | 14 - 15 | 2006 | 6821922 | 96 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |