Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Suz12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001163018.1 | NP_001156490.1 | 5 - 6 | 745 | 98 | |
NM_199196.2 | NP_954666.1 | 6 - 7 | 814 | 98 | ||
GenBank mRNA | AK122213.1 | - | 6 - 7 | 799 | 98 | |
BC064461.1 | - | 6 - 7 | 814 | 6821922 | 98 | |
BC084591.1 | - | 1 - 2 | 85 | 98 | ||
W13031.1 | - | 1 - 2 | 98 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |