Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Utp6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_144826.3 | NP_659075.1 | 1 - 2 | 170 | 155 | |
GenBank mRNA | AF334387.1 | - | 1 - 2 | 497 | 155 | |
AK029151.1 | - | 1 - 2 | 171 | 155 | ||
AK084976.1 | - | 1 - 2 | 163 | 155 | ||
AK164010.1 | - | 1 - 2 | 175 | 155 | ||
AK169785.1 | - | 1 - 2 | 177 | 155 | ||
BC018250.1 | - | 1 - 2 | 134 | 4924739 | 155 | |
BC023920.1 | - | 1 - 2 | 164 | 5323367 | 155 | |
BC024850.1 | - | 1 - 2 | 165 | 5360921 | 155 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus 1700 Probe spans exons |