Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Suz12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001163018.1 | NP_001156490.1 | 11 - 12 | 1591 | 91 | |
NM_199196.2 | NP_954666.1 | 12 - 13 | 1660 | 91 | ||
GenBank mRNA | AK122213.1 | - | 12 - 13 | 1645 | 91 | |
AK135897.1 | - | 6 - 7 | 706 | 91 | ||
AK141264.1 | - | 3 - 4 | 264 | 91 | ||
BC039915.1 | - | 6 - 7 | 736 | 91 | ||
BC051099.1 | - | 3 - 4 | 354 | 91 | ||
BC064461.1 | - | 12 - 13 | 1660 | 6821922 | 91 | |
BC084591.1 | - | 7 - 8 | 931 | 91 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |