Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mbtd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_134012.3 | NP_598773.2 | 10 - 11 | 1366 | 84 | |
GenBank mRNA | AK028503.1 | - | 3 - 4 | 84 | ||
AK033238.1 | - | 5 - 6 | 4239 | 84 | ||
AK141155.1 | - | 10 - 11 | 1361 | 84 | ||
BC020018.1 | - | 6 - 7 | 2646754 | 84 | ||
BC062907.1 | - | 10 - 11 | 1366 | 6813849 | 84 | |
BC064014.1 | - | 12 - 13 | 1470 | 6335954 | 84 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |