Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pgap3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033537.2 | NP_001028709.2 | 1 - 2 | 275 | 74 | |
GenBank mRNA | AK052486.1 | - | 1 - 2 | 192 | 74 | |
AK078974.1 | - | 1 - 2 | 200 | 74 | ||
AK170817.1 | - | 1 - 2 | 190 | 74 | ||
BC035945.1 | - | 1 - 2 | 198 | 74 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |