Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Il31ra | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_139299.2 | NP_647460.2 | 5 - 6 | 593 | 92 | |
GenBank mRNA | AB083111.1 | - | 5 - 6 | 592 | 92 | |
AF486621.1 | - | 2 - 3 | 172 | 92 | ||
AY509150.1 | - | 489 | 92 | |||
AY509151.1 | - | 489 | 92 | |||
BC066164.1 | - | 3 - 4 | 314 | 30475340 | 92 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |