Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ipo11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_029665.3 | NP_083941.2 | 9 - 10 | 994 | 152 | |
GenBank mRNA | AK044487.1 | - | 9 - 10 | 1026 | 152 | |
AK087793.1 | - | 9 - 10 | 1027 | 152 | ||
BC031900.1 | - | 9 - 10 | 930 | 5364208 | 152 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |