Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Smg7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001005507.2 | NP_001005507.1 | 5 - 6 | 598 | 60 | |
NM_001160256.1 | NP_001153728.1 | 5 - 6 | 598 | 60 | ||
NM_001160257.1 | NP_001153729.1 | 5 - 6 | 588 | 60 | ||
GenBank mRNA | AK129101.1 | - | 5 - 6 | 588 | 60 | |
BC082789.1 | - | 5 - 6 | 585 | 30105222 | 60 | |
BC086651.1 | - | 5 - 6 | 598 | 6851591 | 60 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Mus musculus Probe spans exons |