Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Farp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_145519.2 | NP_663494.2 | 9 - 10 | 973 | 107 | |
GenBank mRNA | AK050860.1 | - | 9 - 10 | 933 | 107 | |
AK086699.1 | - | 9 - 10 | 967 | 107 | ||
AK171713.1 | - | 9 - 10 | 999 | 107 | ||
BC009153.1 | - | 9 - 10 | 962 | 2655209 | 107 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |