Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zcchc18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035509.1 | NP_001030586.1 | 1 - 2 | 250 | 79 | |
GenBank mRNA | BC017627.1 | - | 1 - 2 | 111 | 4505033 | 79 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |