Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc35b1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_016752.1 | NP_058032.1 | 3 - 4 | 423 | 61 | |
GenBank mRNA | AK002961.1 | - | 3 - 4 | 417 | 61 | |
AK007711.1 | - | 3 - 4 | 429 | 61 | ||
AK160124.1 | - | 3 - 4 | 489 | 61 | ||
AK183623.1 | - | 2 - 3 | 207 | 61 | ||
AK184790.1 | - | 2 - 3 | 207 | 61 | ||
BC002029.1 | - | 3 - 4 | 404 | 3592141 | 61 | |
D87990.1 | - | 3 - 4 | 423 | 61 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Transporter Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |