Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spryd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033277.3 | NP_001028449.2 | 1 - 2 | 84 | 75 | |
GenBank mRNA | AK146689.1 | - | 1 - 2 | 87 | 75 | |
BC048820.1 | - | 1 - 2 | 61 | 75 | ||
BY739425.1 | - | 1 - 2 | 84 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Transporter Mammalian Gene Collection Mus musculus Probe spans exons |