Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Krt24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_029393.1 | NP_083669.1 | 1 - 2 | 643 | 88 | |
GenBank mRNA | AK010165.1 | - | 1 - 2 | 668 | 88 | |
AK053678.1 | - | 1 - 2 | 389 | 88 | ||
BC129847.1 | - | 1 - 2 | 643 | 40107361 | 88 | |
BC129848.1 | - | 1 - 2 | 643 | 40107363 | 88 | |
GU046702.1 | - | 1 - 2 | 612 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Recommended Assay Mus musculus Probe spans exons |