Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Srrm2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_175229.3 | NP_780438.2 | 11 - 12 | 7848 | 99 | |
GenBank mRNA | AK122248.1 | - | 2 - 3 | 5033 | 99 | |
BC027781.1 | - | 2 - 3 | 1412 | 99 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |