Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Srrm2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_175229.3 | NP_780438.2 | 4 - 5 | 566 | 116 | |
GenBank mRNA | AK090123.1 | - | 4 - 5 | 527 | 116 | |
AK171001.1 | - | 5 - 6 | 839 | 116 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Detect Genomic DNA Mus musculus Probe spans exons |