Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prss34 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_178372.2 | NP_848459.1 | 2 - 3 | 350 | 84 | |
GenBank mRNA | AK081986.1 | - | 2 - 3 | 352 | 84 | |
AY261775.1 | - | 2 - 3 | 350 | 84 | ||
BC109366.2 | - | 2 - 3 | 324 | 84 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Protease Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |