Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1500006006.mm.1 |
NM_023475.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500006006.mm.1 |
NM_023475.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500006006.mm.1 |
TC1500000766.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1500006009.mm.1 |
NM_023475.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500006009.mm.1 |
NM_023475.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500006009.mm.1 |
TC1500000766.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1500006013.mm.1 |
NM_023475.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500006013.mm.1 |
NM_023475.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500006013.mm.1 |
TC1500000766.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1500006007.mm.1 |
NM_023475.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500006007.mm.1 |
NM_023475.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500006007.mm.1 |
TC1500000766.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1500006010.mm.1 |
NM_023475.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500006010.mm.1 |
NM_023475.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500006010.mm.1 |
TC1500000766.mm.1 |
MTA TC |