Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pdia2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081070.1 | NP_001074539.1 | 4 - 5 | 747 | 77 | |
GenBank mRNA | AK007743.1 | - | 4 - 5 | 77 | ||
BC116671.1 | - | 4 - 5 | 713 | 40107256 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |