Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Copz1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019817.1 | NP_062791.1 | 6 - 7 | 394 | 78 | |
GenBank mRNA | AB037370.1 | - | 6 - 7 | 394 | 78 | |
AK003302.1 | - | 6 - 7 | 402 | 78 | ||
AK083824.1 | - | 6 - 7 | 414 | 78 | ||
AK156758.1 | - | 6 - 7 | 413 | 78 | ||
AK167636.1 | - | 6 - 7 | 402 | 78 | ||
AK168382.1 | - | 6 - 7 | 413 | 78 | ||
AK168471.1 | - | 6 - 7 | 413 | 78 | ||
BC002246.1 | - | 6 - 7 | 400 | 78 | ||
BC023207.1 | - | 5 - 6 | 378 | 78 | ||
BC025041.1 | - | 6 - 7 | 400 | 4954031 | 78 | |
BC039920.1 | - | 6 - 7 | 423 | 78 | ||
BC058524.1 | - | 6 - 7 | 393 | 5691652 | 78 | |
BC062203.1 | - | 6 - 7 | 410 | 78 | ||
BC085314.1 | - | 6 - 7 | 415 | 30441470 | 78 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |