Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tfcp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001289603.1 | NP_001276532.1 | 10 - 11 | 1099 | 67 | |
NM_033476.3 | NP_258437.1 | 10 - 11 | 1099 | 67 | ||
GenBank mRNA | AK141248.1 | - | 4 - 5 | 439 | 67 | |
AK146503.1 | - | 10 - 11 | 1100 | 67 | ||
AK147898.1 | - | 9 - 10 | 1308 | 67 | ||
AK158682.1 | - | 10 - 11 | 1019 | 67 | ||
BC022131.1 | - | 10 - 11 | 1072 | 5098336 | 67 | |
M84809.1 | - | 10 - 11 | 1038 | 67 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |