Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rnasel | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011882.2 | NP_036012.1 | 6 - 7 | 2380 | 63 | |
GenBank mRNA | AF281045.1 | - | 4 - 5 | 1924 | 63 | |
AK053125.1 | - | 5 - 6 | 2079 | 63 | ||
AK054205.1 | - | 4 - 5 | 2875 | 63 | ||
AK140815.1 | - | 6 - 7 | 2380 | 63 | ||
AK172300.1 | - | 3 - 4 | 383 | 63 | ||
BC003873.1 | - | 6 - 7 | 2189 | 63 | ||
BC138013.1 | - | 3 - 4 | 1797 | 8861032 | 63 | |
JX443419.1 | - | 3 - 4 | 1761 | 63 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |