Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Plk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_152804.2 | NP_690017.2 | 12 - 13 | 1883 | 95 | |
GenBank mRNA | AK133962.1 | - | 12 - 13 | 1882 | 95 | |
AK160441.1 | - | 1 - 2 | 95 | |||
AK164365.1 | - | 12 - 13 | 1882 | 95 | ||
BC034513.1 | - | 12 - 13 | 1865 | 3156743 | 95 | |
BC035451.1 | - | 12 - 13 | 1865 | 95 | ||
M96163.1 | - | 12 - 13 | 1849 | 95 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Protein Kinase Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |