Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Grm4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013385.2 | NP_001013403.1 | 4 - 5 | 1393 | 76 | |
NM_001291045.1 | NP_001277974.1 | 4 - 5 | 1393 | 76 | ||
GenBank mRNA | BC072635.1 | - | 7 - 8 | 1770 | 76 | |
BC080284.1 | - | 4 - 5 | 1164 | 6402528 | 76 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 G-protein coupled receptor Mammalian Gene Collection Mus musculus Probe spans exons |