Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Srebf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_033218.1 | NP_150087.1 | 1 - 2 | 223 | 64 | |
GenBank mRNA | AF374267.1 | - | 1 - 2 | 76 | 64 | |
AK032847.1 | - | 1 - 2 | 247 | 64 | ||
AK053802.1 | - | 1 - 2 | 242 | 64 | ||
AK155857.1 | - | 1 - 2 | 291 | 64 | ||
AK172219.1 | - | 1 - 2 | 228 | 64 | ||
AK217643.1 | - | 1 - 2 | 140 | 64 | ||
BC014730.1 | - | 1 - 2 | 213 | 64 | ||
BC026556.1 | - | 1 - 2 | 215 | 64 | ||
BC038284.1 | - | 1 - 2 | 214 | 64 | ||
BC048071.1 | - | 1 - 2 | 228 | 64 | ||
BC069989.1 | - | 1 - 2 | 232 | 64 | ||
BQ965109.1 | - | 1 - 2 | 230 | 64 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons |