Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Itga1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033228.3 | NP_001028400.2 | 20 - 21 | 3124 | 131 | |
GenBank mRNA | AK035870.1 | - | 20 - 21 | 3125 | 131 | |
AK040830.1 | - | 11 - 12 | 1548 | 131 |
Set Membership: |
Amplicon length greater than or equal to 101 Mus musculus Probe spans exons |